siRNA Details
| Gene Symbol: LOC101927314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110757.1 | 5 | 1201 | |||
| GenBank mRNA | AK091365.1 | - | 5 | 1150 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433637.2 | - | 1150 | Chr. 6 - 97702281 - 97708103 [+] on Build GRCh38 | ||

LOC101927314
Genomic Map