siRNA Details
Gene Symbol: IPO5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103741.1 | 5 | 576 | |||
NR_103742.1 | 4 | 501 | ||||
GenBank mRNA | AK023040.1 | - | 5 | 555 | ||
AK056739.1 | - | 2 | 1962 | |||
DA183233.1 | - | 4 | 466 | |||
DA723810.1 | - | 4 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000638174.1 | - | 1962 | Chr. - - [-] on Build |
