siRNA Details
| Gene Symbol: LOC158434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132344.1 | 3 | 875 | |||
| GenBank mRNA | AK055173.1 | - | 3 | 876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENSG00000275465
LOC158434| Gene Symbol: LOC158434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132344.1 | 3 | 875 | |||
| GenBank mRNA | AK055173.1 | - | 3 | 876 | ||