siRNA Details
| Gene Symbol: LOC101928514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110837.1 | 2 | 1932 | |||
| GenBank mRNA | AK055500.1 | - | 2 | 1945 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000585863.2 | - | 2 | 1932 | Chr. 17 - 76799044 - 76805953 [+] on Build GRCh38 | |

LOC101928514
Genomic Map