siRNA Details
| Gene Symbol: LINC00462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_051983.1 | 3 | 881 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000425350.1 | - | 2 | 820 | Chr. 13 - 48576974 - 48578088 [-] on Build GRCh38 | |

LINC00462
Genomic Map