siRNA Details
| Gene Symbol: LOC107546764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136406.1 | 2 | 516 | |||
| GenBank mRNA | BX097691.1 | - | 2 | 415 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000565120.1 | - | 2 | 414 | Chr. 17 - 43938363 - 43938959 [+] on Build GRCh38 | |

LOC107546764
Genomic Map