siRNA Details
| Gene Symbol: ARRDC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027435.1 | 3 | 1117 | |||
| GenBank mRNA | BC043415.2 | - | 3 | 1106 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000625713.2 | - | 1115 | Chr. - - [-] on Build | ||

ARRDC3-AS1
Genomic Map