siRNA Details
| Gene Symbol: LOC284933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC048428.1 | - | 3 | 899 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000625694.1 | - | 900 | Chr. - - [-] on Build | ||

LOC284933
Genomic Map