siRNA Details
Gene Symbol: LOC100132147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135764.1 | 2 | 1505 | |||
GenBank mRNA | BC036435.1 | - | 2 | 1508 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000415386.2 | - | 2 | 1502 | Chr. 1 - 16533886 - 16535649 [-] on Build GRCh38 |
