siRNA Details
| Gene Symbol: EML2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130728.1 | 2 | 424 | |||
| GenBank mRNA | BC000856.1 | - | 1 | 145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EML2-AS1| Gene Symbol: EML2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130728.1 | 2 | 424 | |||
| GenBank mRNA | BC000856.1 | - | 1 | 145 | ||