siRNA Details
| Gene Symbol: LOC101929151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135691.1 | 2 | 1962 | |||
| GenBank mRNA | BC040579.1 | - | 2 | 1962 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000630447.1 | - | 1962 | Chr. - - [-] on Build | ||

LOC101929151
Genomic Map