siRNA Details
Gene Symbol: LOC101929151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135691.1 | 2 | 1020 | |||
GenBank mRNA | BC040579.1 | - | 2 | 1020 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000630447.1 | - | 1020 | Chr. - - [-] on Build |
