siRNA Details
Gene Symbol: HCG27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026791.1 | 2 | 1293 | |||
GenBank mRNA | AK097442.1 | - | 2 | 1293 | ||
BC041465.2 | - | 1 | 1172 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HCG27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026791.1 | 2 | 1293 | |||
GenBank mRNA | AK097442.1 | - | 2 | 1293 | ||
BC041465.2 | - | 1 | 1172 |