siRNA Details
| Gene Symbol: C18orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272093.2 | NP_001259022.2 | 2 | 1566 | ||
| GenBank mRNA | AK126293.1 | - | 2 | 1566 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000622985.1 | - | 1566 | Chr. - - [-] on Build | ||

C18orf65
Genomic Map