siRNA Details
| Gene Symbol: LOC100507073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110156.1 | 6 | 639 | |||
| GenBank mRNA | CN360491.1 | - | 6 | 639 | ||
| DA610966.1 | - | 3 | 436 | |||
| DA614343.1 | - | 3 | 447 | |||
| DA616181.1 | - | 3 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507073| Gene Symbol: LOC100507073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110156.1 | 6 | 639 | |||
| GenBank mRNA | CN360491.1 | - | 6 | 639 | ||
| DA610966.1 | - | 3 | 436 | |||
| DA614343.1 | - | 3 | 447 | |||
| DA616181.1 | - | 3 | 444 | |||