siRNA Details
| Gene Symbol: PCBP2-OT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109828.1 | 1 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCBP2-OT1| Gene Symbol: PCBP2-OT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109828.1 | 1 | 100 | |||