siRNA Details
| Gene Symbol: FAM3D-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134853.1 | 1 | 16 | |||
| GenBank mRNA | BG215913.1 | - | 1 | 16 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM3D-AS1| Gene Symbol: FAM3D-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134853.1 | 1 | 16 | |||
| GenBank mRNA | BG215913.1 | - | 1 | 16 | ||