siRNA Details
| Gene Symbol: LINC00629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038998.1 | 3 | 827 | |||
| GenBank mRNA | AK022994.1 | - | 2 | 529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00629| Gene Symbol: LINC00629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038998.1 | 3 | 827 | |||
| GenBank mRNA | AK022994.1 | - | 2 | 529 | ||