siRNA Details
| Gene Symbol: LINC00629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038998.1 | 2 | 271 | |||
| GenBank mRNA | KM886856.1 | - | 2 | 162 | ||
| KM886857.1 | - | 2 | 162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00629| Gene Symbol: LINC00629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038998.1 | 2 | 271 | |||
| GenBank mRNA | KM886856.1 | - | 2 | 162 | ||
| KM886857.1 | - | 2 | 162 | |||