siRNA Details
| Gene Symbol: LINC01370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109936.1 | 3 | 361 | |||
| NR_109937.1 | 3 | 277 | ||||
| GenBank mRNA | BQ417071.1 | - | 2 | 267 | ||
| CK821054.1 | - | 3 | 361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01370| Gene Symbol: LINC01370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109936.1 | 3 | 361 | |||
| NR_109937.1 | 3 | 277 | ||||
| GenBank mRNA | BQ417071.1 | - | 2 | 267 | ||
| CK821054.1 | - | 3 | 361 | |||