siRNA Details
| Gene Symbol: LINC01616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132652.1 | 1 | 2198 | |||
| GenBank mRNA | AI693157.1 | - | 1 | 209 | ||
| AK095844.1 | - | 1 | 2197 | |||
| BE219832.1 | - | 1 | 209 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01616| Gene Symbol: LINC01616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132652.1 | 1 | 2198 | |||
| GenBank mRNA | AI693157.1 | - | 1 | 209 | ||
| AK095844.1 | - | 1 | 2197 | |||
| BE219832.1 | - | 1 | 209 | |||