siRNA Details
| Gene Symbol: LINC01616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132652.1 | 1 | 1296 | |||
| GenBank mRNA | AK095844.1 | - | 1 | 1295 | ||
| BX419799.2 | - | 1 | 677 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX463978.2 | - | 1 | 673 | Chr. 11 - 29980778 - 29981770 [-] on Build GRCh38 | |

LINC01616
Genomic Map