siRNA Details
| Gene Symbol: LINC00160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024351.2 | 1, 2 | 80 | |||
| GenBank mRNA | BX110236.1 | - | 1, 2 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00160| Gene Symbol: LINC00160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024351.2 | 1, 2 | 80 | |||
| GenBank mRNA | BX110236.1 | - | 1, 2 | 80 | ||