siRNA Details
Gene Symbol: LOC102800447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110564.1 | 5 | 560 | |||
GenBank mRNA | BC040863.1 | - | 5 | 559 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102800447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110564.1 | 5 | 560 | |||
GenBank mRNA | BC040863.1 | - | 5 | 559 |