siRNA Details
| Gene Symbol: DBET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121644.1 | 1 | 527 | |||
| GenBank mRNA | JQ639078.1 | - | 1 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DBET| Gene Symbol: DBET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121644.1 | 1 | 527 | |||
| GenBank mRNA | JQ639078.1 | - | 1 | 527 | ||