siRNA Details
| Gene Symbol: LINC00601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073453.1 | 4 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00601| Gene Symbol: LINC00601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073453.1 | 4 | 782 | |||