siRNA Details
| Gene Symbol: PRNCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109833.1 | 1 | 204 | |||
| GenBank mRNA | AB458446.1 | - | 1 | 204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRNCR1| Gene Symbol: PRNCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109833.1 | 1 | 204 | |||
| GenBank mRNA | AB458446.1 | - | 1 | 204 | ||