siRNA Details
| Gene Symbol: SMIM2-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046843.1 | 3 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM2-IT1| Gene Symbol: SMIM2-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046843.1 | 3 | 415 | |||