siRNA Details
| Gene Symbol: LOC102724908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135636.1 | 4 | 2564 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724908| Gene Symbol: LOC102724908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135636.1 | 4 | 2564 | |||