siRNA Details
| Gene Symbol: RNF157-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040017.1 | 7 | 1485 | |||
| GenBank mRNA | AK093500.1 | - | 7 | 1485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF157-AS1| Gene Symbol: RNF157-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040017.1 | 7 | 1485 | |||
| GenBank mRNA | AK093500.1 | - | 7 | 1485 | ||