siRNA Details
Gene Symbol: LOC101928659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135180.1 | 2 | 361 | |||
GenBank mRNA | BQ640484.1 | - | 2 | 358 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135180.1 | 2 | 361 | |||
GenBank mRNA | BQ640484.1 | - | 2 | 358 |