siRNA Details
| Gene Symbol: LDLRAD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040031.1 | 4 | 1858 | |||
| GenBank mRNA | AK098298.1 | - | 4 | 1804 | ||
| BC042085.1 | - | 4 | 1843 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LDLRAD4-AS1| Gene Symbol: LDLRAD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040031.1 | 4 | 1858 | |||
| GenBank mRNA | AK098298.1 | - | 4 | 1804 | ||
| BC042085.1 | - | 4 | 1843 | |||