siRNA Details
| Gene Symbol: LINC00461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024383.1 | 4 | 1397 | |||
| NR_024384.1 | 3 | 1599 | ||||
| GenBank mRNA | AK055372.1 | - | 1 | 376 | ||
| AK091356.1 | - | 4 | 2212 | |||
| BC036480.2 | - | 4 | 1420 | |||
| BC047100.1 | - | 3 | 1408 | |||
| BC048008.1 | - | 3 | 1408 | |||

LINC00461
Genomic Map