siRNA Details
Gene Symbol: LINC00461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024383.1 | 4 | 748 | |||
NR_024384.1 | 3 | 950 | ||||
GenBank mRNA | AK091356.1 | - | 4 | 1563 | ||
AK308694.1 | - | 3 | 949 | |||
AK311230.1 | - | 3 | 940 | |||
BC036480.2 | - | 4 | 770 | |||
BC047100.1 | - | 3 | 758 | |||
BC048008.1 | - | 3 | 758 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX460025.2 | - | 4 | 762 | Chr. 5 - 88666832 - 88684768 [-] on Build GRCh38 |
