siRNA Details
| Gene Symbol: ROR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110666.1 | 6 | 1097 | |||
| GenBank mRNA | BC040909.1 | - | 6 | 1097 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROR1-AS1| Gene Symbol: ROR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110666.1 | 6 | 1097 | |||
| GenBank mRNA | BC040909.1 | - | 6 | 1097 | ||