siRNA Details
| Gene Symbol: NRG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120666.1 | 1, 2 | 184 | |||
| GenBank mRNA | AA383013.1 | - | 1, 2 | 119 | ||
| CR737767.1 | - | 1, 2 | 119 | |||
| DB052745.1 | - | 1, 2 | 183 | |||
| DB082602.1 | - | 1, 2 | 185 | |||

NRG3-AS1
Genomic Map