siRNA Details
| Gene Symbol: NRG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120666.1 | 3 | 363 | |||
| GenBank mRNA | AA383013.1 | - | 3 | 298 | ||
| CF272718.1 | - | 2 | 141 | |||
| CR737767.1 | - | 3 | 298 | |||
| DB052745.1 | - | 3 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NRG3-AS1| Gene Symbol: NRG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120666.1 | 3 | 363 | |||
| GenBank mRNA | AA383013.1 | - | 3 | 298 | ||
| CF272718.1 | - | 2 | 141 | |||
| CR737767.1 | - | 3 | 298 | |||
| DB052745.1 | - | 3 | 362 | |||