siRNA Details
Gene Symbol: LINC00690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103843.1 | 3 | 541 | |||
GenBank mRNA | BC034913.1 | - | 3 | 544 | ||
BX100558.1 | - | 3 | 541 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103843.1 | 3 | 541 | |||
GenBank mRNA | BC034913.1 | - | 3 | 544 | ||
BX100558.1 | - | 3 | 541 |