siRNA Details
| Gene Symbol: NRSN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109990.1 | 3 | 502 | |||
| GenBank mRNA | BX104512.1 | - | 3 | 500 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AW970418.1 | - | 3 | 485 | Chr. 20 - 320427 - 325228 [-] on Build GRCh38 | |

NRSN2-AS1
Genomic Map