siRNA Details
| Gene Symbol: LOC284661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027088.1 | 2, 3 | 620 | |||
| GenBank mRNA | AK093498.1 | - | 2, 3 | 620 | ||
| BC128213.1 | - | 1, 2 | 32 | |||
| BC128214.1 | - | 1, 2 | 33 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB043752.1 | - | 2, 3 | 228 | Chr. 1 - 4412161 - 4422877 [+] on Build GRCh38 | |

LOC284661
Genomic Map