siRNA Details
| Gene Symbol: LOC102724392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134268.1 | 3 | 2199 | |||
| GenBank mRNA | AK123868.1 | - | 2 | 2369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724392| Gene Symbol: LOC102724392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134268.1 | 3 | 2199 | |||
| GenBank mRNA | AK123868.1 | - | 2 | 2369 | ||