siRNA Details
| Gene Symbol: LOC102724392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134268.1 | 3 | 736 | |||
| GenBank mRNA | AK123868.1 | - | 2 | 906 | ||
| AK310013.1 | - | 3 | 736 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF141636.1 | - | 2 | 301 | Chr. 5 - 71353470 - 71445734 [-] on Build GRCh38 | |

LOC102724392
Genomic Map