siRNA Details
Gene Symbol: LINC01219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126400.1 | 3 | 590 | |||
GenBank mRNA | BX092271.1 | - | 3 | 592 |
Gene Symbol: HOTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293171.2 | NP_001280100.1 | 1 | 930 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR740877.1 | - | 3 | 591 | Chr. 11 - 1991097 - 1993462 [+] on Build GRCh38 |
