siRNA Details
Gene Symbol: LOC101927815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125426.1 | 3 | 558 | |||
GenBank mRNA | AK128880.1 | - | 3 | 524 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX095510.1 | - | 3 | 480 | Chr. 8 - 2696557 - 2728373 [-] on Build GRCh38 |
