siRNA Details
| Gene Symbol: LOC105371592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136412.1 | 2 | 403 | |||
| GenBank mRNA | BU581060.1 | - | 2 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371592| Gene Symbol: LOC105371592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136412.1 | 2 | 403 | |||
| GenBank mRNA | BU581060.1 | - | 2 | 403 | ||