siRNA Details
| Gene Symbol: C20orf187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX281604.1 | - | 3, 4 | 318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf187| Gene Symbol: C20orf187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX281604.1 | - | 3, 4 | 318 | ||