siRNA Details
Gene Symbol: LINC00905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024335.2 | 2 | 259 | |||
GenBank mRNA | AK058057.1 | - | 2 | 218 | ||
BC069223.1 | - | 2 | 191 | |||
BC104221.2 | - | 1 | 156 | |||
DB461577.1 | - | 2 | 259 |
