siRNA Details
Gene Symbol: LINC00905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024335.2 | 5 | 1474 | |||
NR_110321.1 | 5 | 1184 | ||||
GenBank mRNA | AK058057.1 | - | 5 | 1432 | ||
BC031284.1 | - | 5 | 1183 | |||
BC069223.1 | - | 5 | 1405 |
