siRNA Details
| Gene Symbol: LOC284898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318329.1 | NP_001305258.1 | 2 | 1193 | ||
| GenBank mRNA | AK055980.1 | - | 2 | 973 | ||
| BC036876.1 | - | 2 | 1180 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284898| Gene Symbol: LOC284898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318329.1 | NP_001305258.1 | 2 | 1193 | ||
| GenBank mRNA | AK055980.1 | - | 2 | 973 | ||
| BC036876.1 | - | 2 | 1180 | |||