siRNA Details
| Gene Symbol: LOC100506990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040091.1 | 1 | 530 | |||
| NR_040092.1 | 1 | 530 | ||||
| GenBank mRNA | AK095981.1 | - | 1 | 179 | ||
| AK129982.1 | - | 1 | 426 | |||
| AL539380.3 | - | 1 | 529 | |||
| BC037934.1 | - | 1 | 413 | |||

LOC100506990
Genomic Map